Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,6-Dibromo-2-fluoropyridine ≥97% research grade

Supplier: AOB CHEM USA
Warning

53318-1G 53318-25G 53318-500MG 53318-5G
76917-006EA 304.48 USD InStock
76917-006 76917-008 76917-010 76917-012
3,6-Dibromo-2-fluoropyridine ≥97% research grade
3,6-Dibromo-2-fluoropyridine


Formula: C₅H₂Br₂FN
MW: 254.882 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18258101
CAS Number: 1256820-70-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty