Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hydroxydecanoic acid ≥95% research grade

Supplier: AOB CHEM USA
Warning

55425-1G 55425-25G 55425-500MG 55425-5G
76917-194EA 292.81 USD InStock
76917-194 76917-196 76917-198 76917-200
3-Hydroxydecanoic acid ≥95% research grade
3-Hydroxydecanoic acid


Formula: C₁₀H₂₀O₃
MW: 188.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00133275
CAS Number: 14292-26-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty