Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Cyclopropyl-2,6-difluorobenzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

56428-1G 56428-500MG 56428-5G
76917-262EA 558.04 USD InStock
76917-262 76917-264 76917-266
3-Cyclopropyl-2,6-difluorobenzoic acid ≥97% research grade
3-Cyclopropyl-2,6-difluorobenzoic acid


Formula: C₁₀H₈F₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD34530521
CAS Number: 2734775-86-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty