Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-fluoro-5-formylbenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

61338-1G 61338-25G 61338-500MG 61338-5G
76917-522EA 203.7 USD InStock
76917-522 76917-524 76917-526 76917-528
3-Bromo-2-fluoro-5-formylbenzoic acid ≥97%
3-Bromo-2-fluoro-5-formylbenzoic acid


Formula: C₈H₄BrFO₃
Storage Temperature: Ambient
CAS Number: 2387232-88-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty