Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methyl-[1,1'-biphenyl]-4-carboxylic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

65252-1G 65252-25G 65252-500MG 65252-5G
76917-748EA 248.26 USD InStock
76917-748 76917-750 76917-752 76917-754
3-Methyl-[1,1'-biphenyl]-4-carboxylic acid ≥97% research grade
3-Methyl-[1,1'-biphenyl]-4-carboxylic acid


Formula: C₁₄H₁₂O₂
Storage Temperature: Ambient
CAS Number: 69240-50-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty