Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-chloro-1-(methylsulfonyl)benzene ≥97%

Supplier: AOB CHEM USA
Warning

74424-1G 74424-25G 74424-5G
76917-946EA 95.49 USD InStock
76917-946 76917-948 76917-950
4-Bromo-2-chloro-1-(methylsulfonyl)benzene ≥97%
4-Bromo-2-chloro-1-(methylsulfonyl)benzene
Precaución: for Research Use Only


Formula: C₇H₆BrClO₂S
MW: 269.543 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26131172
CAS Number: 648905-09-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty