Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-(methylthio)benzonitrile ≥97% research grade

Supplier: AOB CHEM USA
Warning

75983-1G 75983-500MG 75983-5G
76917-992EA 450.89 USD InStock
76917-992 76917-994 76917-996
3-Bromo-5-(methylthio)benzonitrile ≥97% research grade
3-Bromo-5-(methylthio)benzonitrile


Formula: C₈H₆BrNS
Storage Temperature: Ambient
MDL Number: MFCD22488915
CAS Number: 1595930-26-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty