Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-chloro-6-fluoropyridine ≥97%

Supplier: AOB CHEM USA
Warning

90007-1G 90007-25G 90007-500MG 90007-5G
76918-180EA 217.49 USD InStock
76918-180 76918-182 76918-184 76918-186
3-Bromo-2-chloro-6-fluoropyridine ≥97%
3-Bromo-2-chloro-6-fluoropyridine


Formula: C₅H₂BrClFN
Storage Temperature: Ambient
MDL Number: MFCD18257575
CAS Number: 1211578-75-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty