Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(1,1-Difluoroethyl)benzaldehyde ≥97%

Supplier: AOB CHEM USA
Warning

90569-1G 90569-25G 90569-500MG 90569-5G
76918-196EA 393.6 USD InStock
76918-196 76918-198 76918-200 76918-202
4-(1,1-Difluoroethyl)benzaldehyde ≥97%
4-(1,1-Difluoroethyl)benzaldehyde


Formula: C₉H₈F₂O
Storage Temperature: Ambient
MDL Number: MFCD17015610
CAS Number: 55805-22-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty