Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-6-fluoro-3-proproxyphenylboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

11001-1G 11001-25G 11001-5G
76894-486EA 64.72 USD InStock
76894-486 76894-488 76894-490
2-Bromo-6-fluoro-3-proproxyphenylboronic acid ≥97% research grade
2-Bromo-6-fluoro-3-proproxyphenylboronic acid


Formula: C₉H₁₁BBrFO₃
Storage Temperature: Refrigerator
MDL Number: MFCD06411369
CAS Number: 849052-20-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty