Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-6-methoxypyridine-3-boronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

11631-1G 11631-25G 11631-500MG 11631-5G
76894-634EA 135.8 USD InStock
76894-634 76894-636 76894-638 76894-640
2-Methyl-6-methoxypyridine-3-boronic acid ≥97% research grade
2-Methyl-6-methoxypyridine-3-boronic acid


Formula: C₇H₁₀BNO₃
MW: 166.97 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05662386
CAS Number: 459856-12-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty