Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxy-6-(trifluoromethyl)nicotinic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

121592-1G 121592-25G 121592-500MG 121592-5G
76894-806EA 425.43 USD InStock
76894-806 76894-808 76894-810 76894-812
2-Methoxy-6-(trifluoromethyl)nicotinic acid ≥97% research grade
2-Methoxy-6-(trifluoromethyl)nicotinic acid


Formula: C₈H₆F₃NO₃
MW: 221.133 g/mol
Storage Temperature: Refrigerator
CAS Number: 916160-41-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty