Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-1-ethoxy-4-methybenzene ≥95% research grade

Supplier: AOB CHEM USA
Warning

12767-25G 12767-5G
76894-898EA 346.92 USD InStock
76894-898 76894-900
2-Bromo-1-ethoxy-4-methybenzene ≥95% research grade
2-Bromo-1-ethoxy-4-methybenzene


Formula: C₉H₁₁BrO
MW: 215.087 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11156052
CAS Number: 103260-55-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty