Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2,6-dimethylphenylboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

12770-1G 12770-25G 12770-5G
76894-908EA 114.58 USD InStock
76894-908 76894-910 76894-912
4-Chloro-2,6-dimethylphenylboronic acid ≥97% research grade
4-Chloro-2,6-dimethylphenylboronic acid


Formula: C₈H₁₀BClO₂
Storage Temperature: Ambient
MDL Number: MFCD10566594
CAS Number: 1027045-31-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty