Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Trifluoromethoxy)salicylic acid ≥97%

Supplier: AOB CHEM USA
Warning

12775-1G 12775-25G 12775-5G
76894-920EA 62.6 USD InStock
76894-920 76894-922 76894-924
5-(Trifluoromethoxy)salicylic acid ≥97%
5-(Trifluoromethoxy)salicylic acid
Precaución: for Research Use Only


Formula: C₈H₅F₃O₄
MW: 222.12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01091003
CAS Number: 129644-57-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty