Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dibromopyridine-3-boronicacid ≥97%

Supplier: AOB CHEM USA
Warning

14051-1G 14051-25G 14051-500MG 14051-5G
76895-220EA 281.14 USD InStock
76895-220 76895-222 76895-224 76895-226
2,6-Dibromopyridine-3-boronicacid ≥97%
2,6-Dibromopyridine-3-boronicacid


Formula: C₅H₄BBr₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD11865286
CAS Number: 1256355-52-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty