Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-3-formyl-4-picoline ≥97% research grade

Supplier: AOB CHEM USA
Warning

14141-1G 14141-25G 14141-500MG 14141-5G
76895-292EA 146.41 USD InStock
76895-292 76895-294 76895-296 76895-298
2-Fluoro-3-formyl-4-picoline ≥97% research grade
2-Fluoro-3-formyl-4-picoline


Formula: C₇H₆FNO
MW: 139.127 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08277290
CAS Number: 609771-39-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty