Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-3-methylpyridine-5-carboxylic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

14191-1G 14191-25G 14191-5G
76895-340EA 76.39 USD InStock
76895-340 76895-342 76895-344
2-Bromo-3-methylpyridine-5-carboxylic acid ≥97% research grade
2-Bromo-3-methylpyridine-5-carboxylic acid


Formula: C₇H₆BrNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD11044925
CAS Number: 901300-51-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty