Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-5-isopropoxytoluene 95%

Supplier: AOB CHEM USA
Warning

14542-1G 14542-25G 14542-5G 14542-500MG
76895-556EA 109.28 USD InStock
76895-556 76895-558 76895-560 77644-628
2-Bromo-5-isopropoxytoluene 95%
2-Bromo-5-isopropoxytoluene


Formula: C₁₀H₁₃BrO
Storage Temperature: Refrigerator
MDL Number: MFCD18917791
CAS Number: 86786-24-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty