Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-(trifluoromethoxy)anisole ≥95%

Supplier: AOB CHEM USA
Warning

15449-1G 15449-25G 15449-500MG 15449-5G
76895-940EA 183.54 USD InStock
76895-940 76895-942 76895-944 76895-946
2-Bromo-4-(trifluoromethoxy)anisole ≥95%
2-Bromo-4-(trifluoromethoxy)anisole
Precaución: for Research Use Only


Formula: C₈H₆BrF₃O₂
MW: 271.031 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04973759
CAS Number: 200956-14-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty