Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Dimethylamino)-5-pyrimidineboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

16116-25G 16116-5G
76896-352EA 559.1 USD InStock
76896-352 76896-354
2-(Dimethylamino)-5-pyrimidineboronic acid ≥97%
2-(Dimethylamino)-5-pyrimidineboronic acid
Precaución: for Research Use Only


Formula: C₆H₁₀BN₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD10696933
CAS Number: 756817-82-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty