Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dibromo-4-fluoro-1-propoxybenzene ≥97%

Supplier: AOB CHEM USA
Warning

16522-25G 16522-5G
76896-480EA 346.92 USD InStock
76896-480 76896-482
2,6-Dibromo-4-fluoro-1-propoxybenzene ≥97%
2,6-Dibromo-4-fluoro-1-propoxybenzene


Formula: C₉H₉Br₂FO
Storage Temperature: Ambient
MDL Number: MFCD17011973
CAS Number: 1242070-99-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty