Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-ethyl-1-fluorobenzene ≥97% research grade

Supplier: AOB CHEM USA
Warning

16548-1G 16548-25G 16548-500MG 16548-5G
76896-488EA 126.25 USD InStock
76896-488 76896-490 76896-492 76896-494
2-Bromo-4-ethyl-1-fluorobenzene ≥97% research grade
2-Bromo-4-ethyl-1-fluorobenzene


Formula: C₈H₈BrF
MW: 203.052 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11846040
CAS Number: 891843-33-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty