Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-4-methyl-3-pyridineboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

16704-1G 16704-25G 16704-500MG 16704-5G
76896-506EA 409.51 USD InStock
76896-506 76896-508 76896-510 76896-512
2-Fluoro-4-methyl-3-pyridineboronic acid ≥97%
2-Fluoro-4-methyl-3-pyridineboronic acid
Precaución: for Research Use Only


Formula: C₆H₇BFNO₂
MW: 154.940 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07374980
CAS Number: 1029654-30-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty