Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Methoxy-4,6-bis(trifluoromethyl)phenyl)boronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

17130-1G 17130-25G 17130-500MG 17130-5G
76896-728EA 126.25 USD InStock
76896-728 76896-730 76896-732 76896-734
(2-Methoxy-4,6-bis(trifluoromethyl)phenyl)boronic acid ≥97% research grade
(2-Methoxy-4,6-bis(trifluoromethyl)phenyl)boronic acid


Formula: C₉H₇BF₆O₃
MW: 287.95 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27756648
CAS Number: 1067228-89-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty