Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-5-pyrimidineboronic acid pinacol ester ≥97% research grade

Supplier: AOB CHEM USA
Warning

18038-25G 18038-5G
76897-112EA 386.17 USD InStock
76897-112 76897-114
2-Methyl-5-pyrimidineboronic acid pinacol ester ≥97% research grade
2-Methyl-5-pyrimidineboronic acid pinacol ester


Formula: C₁₁H₁₇N₂O₂B
Storage Temperature: Ambient
MDL Number: MFCD12032615
CAS Number: 1052686-67-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty