Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-5-(phenylcarbamoyl)phenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

18117-1G 18117-25G 18117-500MG 18117-5G
76897-120EA 177.18 USD InStock
76897-120 76897-122 76897-124 76897-126
2-Fluoro-5-(phenylcarbamoyl)phenylboronic acid ≥97%
2-Fluoro-5-(phenylcarbamoyl)phenylboronic acid
Precaución: for Research Use Only


Formula: C₁₃H₁₁BFNO₃
MW: 259.0400 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08235095
CAS Number: 874289-41-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty