Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Difluoro-3-nitrophenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

18584-25G 18584-5G
76897-256EA 552.73 USD InStock
76897-256 76897-258
2,6-Difluoro-3-nitrophenylboronic acid ≥97%
2,6-Difluoro-3-nitrophenylboronic acid
Precaución: for Research Use Only


Formula: C₆H₄BF₂NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD12025957
CAS Number: 1150114-28-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty