Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Difluoro-4-formylbenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

20123-1G 20123-25G 20123-5G 20123-500MG
76897-792EA 94.43 USD InStock
76897-792 76897-794 76897-796 77649-206
2,6-Difluoro-4-formylbenzoic acid ≥97%
2,6-Difluoro-4-formylbenzoic acid


Formula: C₈H₄F₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD17011991
CAS Number: 1242070-98-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty