Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Benzyloxy-3-bromopyridine ≥97% research grade

Supplier: AOB CHEM USA
Warning

20173-25G 20173-5G
76897-812EA 477.41 USD InStock
76897-812 76897-814
2-Benzyloxy-3-bromopyridine ≥97% research grade
2-Benzyloxy-3-bromopyridine


Formula: C₁₂H₁₀BrNO
MW: 264.117 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09037467
CAS Number: 52200-49-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty