Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-5-(methylthio)phenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

20280-1G 20280-25G 20280-500MG 20280-5G
76897-856EA 337.37 USD InStock
76897-856 76897-858 76897-860 76897-862
2-Fluoro-5-(methylthio)phenylboronic acid ≥97%
2-Fluoro-5-(methylthio)phenylboronic acid


Formula: C₇H₈BFO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD29088118
CAS Number: 2121512-51-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty