Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-fluoro-6-iodobenzoate ≥97% research grade

Supplier: AOB CHEM USA
Warning

20992-1G 20992-25G 20992-500MG 20992-5G
76898-266EA 228.1 USD InStock
76898-266 76898-268 76898-270 76898-272
Methyl 2-fluoro-6-iodobenzoate ≥97% research grade
Methyl 2-fluoro-6-iodobenzoate


Formula: C₈H₆FIO₂
MW: 280.0349 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12025052
CAS Number: 146014-66-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty