Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dichloro-4-methylboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

21150-1G 21150-25G 21150-500MG 21150-5G
76898-302EA 385.11 USD InStock
76898-302 76898-304 76898-306 76898-308
2,5-Dichloro-4-methylboronic acid ≥97% research grade
2,5-Dichloro-4-methylboronic acid


Formula: C₇H₇BCl₂O₂
MW: 204.846 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09998847
CAS Number: 2246725-44-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty