Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Difluoro-3-propoxyphenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

21240-25G 21240-5G
76898-330EA 580.32 USD InStock
76898-330 76898-332
2,6-Difluoro-3-propoxyphenylboronic acid ≥97%
2,6-Difluoro-3-propoxyphenylboronic acid
Precaución: for Research Use Only


Formula: C₉H₁₁BF₂O₃
MW: 215.99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07369752
CAS Number: 849062-14-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty