Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-6-chlorobenzyl alcohol ≥97% research grade

Supplier: AOB CHEM USA
Warning

21469-1G 21469-25G 21469-5G
76898-398EA 79.57 USD InStock
76898-398 76898-400 76898-402
2-Bromo-6-chlorobenzyl alcohol ≥97% research grade
2-Bromo-6-chlorobenzyl alcohol


Formula: C₇H₆BrClO
Storage Temperature: Ambient
MDL Number: MFCD18390310
CAS Number: 1242822-57-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty