Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Benzyloxy-3-chlorophenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

21491-1G 21491-25G 21491-5G
76898-408EA 99.73 USD InStock
76898-408 76898-410 76898-412
2-Benzyloxy-3-chlorophenylboronic acid ≥97%
2-Benzyloxy-3-chlorophenylboronic acid
Precaución: for Research Use Only


Formula: C₁₃H₁₂BClO₃
MW: 262.5000 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13195644
CAS Number: 1217500-57-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty