Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-methoxymethylphenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

21601-1G 21601-25G 21601-500MG 21601-5G
76898-442EA 290.69 USD InStock
76898-442 76898-444 76898-446 76898-448
2-Chloro-5-methoxymethylphenylboronic acid ≥97%
2-Chloro-5-methoxymethylphenylboronic acid


Formula: C₈H₁₀BClO₃
Storage Temperature: Refrigerator
MDL Number: MFCD19105382
CAS Number: 1451392-23-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty