Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-3-hydroxymethylphenylboronic acid ≥95%

Supplier: AOB CHEM USA
Warning

21836-1G 21836-25G 21836-500MG 21836-5G
76898-516EA 135.8 USD InStock
76898-516 76898-518 76898-520 76898-522
2-Fluoro-3-hydroxymethylphenylboronic acid ≥95%
2-Fluoro-3-hydroxymethylphenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₈BFO₃
Storage Temperature: Refrigerator
MDL Number: MFCD17926485
CAS Number: 1256355-08-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty