Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-(trifluoromethoxy)benzonic acid ≥97%

Supplier: AOB CHEM USA
Warning

22140-1G 22140-25G 22140-500MG 22140-5G
76898-610EA 188.85 USD InStock
76898-610 76898-612 76898-614 76898-616
2-Bromo-4-(trifluoromethoxy)benzonic acid ≥97%
2-Bromo-4-(trifluoromethoxy)benzonic acid


Formula: C₈H₄BrF₃O₃
Storage Temperature: Refrigerator
MDL Number: MFCD13185876
CAS Number: 1138331-74-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty