Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-3,6-difluorobenzeneboronic acid ≥95%

Supplier: AOB CHEM USA
Warning

22284-1G 22284-25G 22284-500MG 22284-5G
76898-634EA 292.81 USD InStock
76898-634 76898-636 76898-638 76898-640
2-Bromo-3,6-difluorobenzeneboronic acid ≥95%
2-Bromo-3,6-difluorobenzeneboronic acid
Precaución: for Research Use Only


Formula: C₆H₄BBrF₂O₂
MW: 236.807 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12026702
CAS Number: 1260757-41-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty