Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxymethyl-5-(trifluoromethyl)phenylboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

22539-1G 22539-25G 22539-500MG 22539-5G
76898-738EA 263.11 USD InStock
76898-738 76898-740 76898-742 76898-744
2-Hydroxymethyl-5-(trifluoromethyl)phenylboronic acid ≥97% research grade
2-Hydroxymethyl-5-(trifluoromethyl)phenylboronic acid
Precaución: for Research Use Only


Formula: C₈H₈BF₃O₃
MW: 219.954 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20526380
CAS Number: 174671-50-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty