Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Ethoxycarbonyl)phenylboronic acid pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

22945-25G 22945-5G
76898-936EA 204.76 USD InStock
76898-936 76898-938
2-(Ethoxycarbonyl)phenylboronic acid pinacol ester ≥97%
2-(Ethoxycarbonyl)phenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₅H₂₁BO₄
Storage Temperature: Ambient
MDL Number: MFCD03453660
CAS Number: 269409-99-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty