Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dibromo-4-(trifluoromethyl)phenol ≥97%

Supplier: AOB CHEM USA
Warning

22972-1G 22972-25G 22972-500MG 22972-5G
76898-948EA 144.29 USD InStock
76898-948 76898-950 76898-952 76898-954
2,6-Dibromo-4-(trifluoromethyl)phenol ≥97%
2,6-Dibromo-4-(trifluoromethyl)phenol
Precaución: for Research Use Only


Formula: C₇H₃Br₂F₃O
Storage Temperature: Refrigerator
MDL Number: MFCD09261146
CAS Number: 35852-57-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty