Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Difluoro-4-hydroxybenzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

23037-1G 23037-25G 23037-5G
76898-986EA 88.06 USD InStock
76898-986 76898-988 76898-990
2,5-Difluoro-4-hydroxybenzoic acid ≥97% research grade
2,5-Difluoro-4-hydroxybenzoic acid


Formula: C₇H₄F₂O₃
MW: 174.102 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04112525
CAS Number: 146781-23-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty