Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Difluoro-3-(1-methylethoxy)benzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

23387-1G 23387-25G 23387-500MG 23387-5G
76899-018EA 202.64 USD InStock
76899-018 76899-020 76899-022 76899-024
2,4-Difluoro-3-(1-methylethoxy)benzoic acid ≥97%
2,4-Difluoro-3-(1-methylethoxy)benzoic acid


Formula: C₁₀H₁₀F₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD22056760
CAS Number: 1219020-69-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty