Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-3-iodo-5-methoxypyridine ≥97%

Supplier: AOB CHEM USA
Warning

23504-1G 23504-25G 23504-500MG 23504-5G
76899-080EA 171.87 USD InStock
76899-080 76899-082 76899-084 76899-086
2-Fluoro-3-iodo-5-methoxypyridine ≥97%
2-Fluoro-3-iodo-5-methoxypyridine


Formula: C₆H₅FINO
Storage Temperature: Refrigerator
MDL Number: MFCD16611134
CAS Number: 1227579-57-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty