Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Bromo-5-methoxyphenyl)methanol ≥97%

Supplier: AOB CHEM USA
Warning

24188-25G 24188-5G
76899-382EA 279.02 USD InStock
76899-382 76899-384
(2-Bromo-5-methoxyphenyl)methanol ≥97%
(2-Bromo-5-methoxyphenyl)methanol


Formula: C₈H₉BrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD03695463
CAS Number: 150192-39-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty