Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Cyclopropyl-4-(trifluoromethoxy)aniline ≥97% research grade

Supplier: AOB CHEM USA
Warning

25622-1G 25622-25G 25622-500MG 25622-5G
76900-040EA 409.51 USD InStock
76900-040 76900-042 76900-044 76900-046
2-Cyclopropyl-4-(trifluoromethoxy)aniline ≥97% research grade
2-Cyclopropyl-4-(trifluoromethoxy)aniline


Formula: C₁₀H₁₀F₃NO
MW: 217.188 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23713135
CAS Number: 1428234-58-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty