Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Difluoro-3-(methylthio)benzyl alcohol ≥97% research grade

Supplier: AOB CHEM USA
Warning

25803-1G 25803-25G 25803-500MG 25803-5G
76900-122EA 320.4 USD InStock
76900-122 76900-124 76900-126 76900-128
2,6-Difluoro-3-(methylthio)benzyl alcohol ≥97% research grade
2,6-Difluoro-3-(methylthio)benzyl alcohol


Formula: C₈H₈F₂OS
Storage Temperature: Ambient
MDL Number: MFCD23713148
CAS Number: 1428234-82-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty