Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-4-methoxypyridine-3-boronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

26140-1G 26140-500MG 26140-5G
76900-252EA 485.9 USD InStock
76900-252 76900-254 76900-256
2-Fluoro-4-methoxypyridine-3-boronic acid ≥97% research grade
2-Fluoro-4-methoxypyridine-3-boronic acid


Formula: C₆H₇BFNO₃
MW: 170.934 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18258142
CAS Number: 2121513-35-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty